Search
Search

ACSS2| Gene Symbol: ACSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710791.3 | XP_002710837.1 | 18 - 19 | 2106 | 56 | |
| XM_002710792.3 | XP_002710838.1 | 17 - 18 | 2065 | 56 | ||
Set Membership: |
|