Search
Search

MYBBP1A| Gene Symbol: MYBBP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270864.2 | XP_008269086.1 | 5 - 6 | 647 | 54 | |
| XM_008270865.2 | XP_008269087.1 | 5 - 6 | 647 | 54 | ||
| XM_008270866.2 | XP_008269088.1 | 5 - 6 | 647 | 54 | ||
| XM_017348928.1 | XP_017204417.1 | 3 - 4 | 268 | 54 | ||
Set Membership: |
|