Search
Search

CA5A| Gene Symbol: CA5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017339451.1 | XP_017194940.1 | 490 | 82 | ||
| XM_017339452.1 | XP_017194941.1 | 492 | 82 | |||
| XM_017339453.1 | XP_017194942.1 | 480 | 82 | |||
| XM_017339455.1 | XP_017194944.1 | 287 | 82 | |||
Set Membership: |
|