Search
Search

COPS5| Gene Symbol: COPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710454.3 | XP_002710500.1 | 4 - 5 | 713 | 109 | |
| XM_008255620.2 | XP_008253842.1 | 4 - 5 | 554 | 109 | ||
Set Membership: |
|