Search
Search

NRAP| Gene Symbol: NRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718751.3 | XP_002718797.1 | 32 - 33 | 3681 | 60 | |
| XM_002718752.3 | XP_002718798.1 | 31 - 32 | 3576 | 60 | ||
| XM_008270528.2 | XP_008268750.1 | 32 - 33 | 3862 | 60 | ||
| XM_017348567.1 | XP_017204056.1 | 33 - 34 | 3865 | 60 | ||
| XM_017348568.1 | XP_017204057.1 | 33 - 34 | 3862 | 60 | ||
| XM_017348569.1 | XP_017204058.1 | 32 - 33 | 3760 | 60 | ||
| XM_017348570.1 | XP_017204059.1 | 32 - 33 | 3757 | 60 | ||
Set Membership: |
|