Search
Search

ARHGAP29| Gene Symbol: ARHGAP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715850.3 | XP_002715896.1 | 13 - 14 | 1555 | 61 | |
| XM_008264860.2 | XP_008263082.1 | 13 - 14 | 1616 | 61 | ||
| XM_017346068.1 | XP_017201557.1 | 13 - 14 | 1618 | 61 | ||
Set Membership: |
|