Search
Search

RGL2| Gene Symbol: RGL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714446.2 | XP_002714492.1 | 17 - 18 | 2258 | 92 | |
| XM_008262780.1 | XP_008261002.1 | 17 - 18 | 2261 | 92 | ||
| XM_008262781.1 | XP_008261003.1 | 17 - 18 | 1976 | 92 | ||
Set Membership: |
|