Search
Search

SUPT20H| Gene Symbol: SUPT20H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720594.3 | XP_002720640.2 | 1832 | 72 | ||
| XM_008273562.2 | XP_008271784.1 | 1650 | 72 | |||
| XM_008273563.2 | XP_008271785.1 | 1571 | 72 | |||
| XM_008273564.2 | XP_008271786.1 | 1832 | 72 | |||
| XM_008273566.2 | XP_008271788.1 | 1831 | 72 | |||
| XM_008273568.2 | XP_008271790.1 | 1760 | 72 | |||
| XM_017350158.1 | XP_017205647.1 | 1835 | 72 | |||
| XM_017350159.1 | XP_017205648.1 | 1702 | 72 | |||
| XM_017350160.1 | XP_017205649.1 | 1620 | 72 | |||
| XM_017350161.1 | XP_017205650.1 | 1833 | 72 | |||
| XM_017350162.1 | XP_017205651.1 | 1808 | 72 | |||
| XM_017350163.1 | XP_017205652.1 | 1834 | 72 | |||
| XM_017350164.1 | XP_017205653.1 | 1835 | 72 | |||
| XM_017350165.1 | XP_017205654.1 | 1834 | 72 | |||
| XM_017350166.1 | XP_017205655.1 | 1831 | 72 | |||
Set Membership: |
|