Search
Search

XPO4| Gene Symbol: XPO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712804.3 | XP_002712850.1 | 9 - 10 | 1314 | 88 | |
| XM_008259924.2 | XP_008258146.1 | 10 - 11 | 1420 | 88 | ||
Set Membership: |
|