Search
Search

TRIM7| Gene Symbol: TRIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713882.2 | XP_002713928.1 | 1 - 2 | 821 | 71 | |
| XM_008262006.1 | XP_008260228.1 | 1 - 2 | 821 | 71 | ||
| XM_017344641.1 | XP_017200130.1 | 1 - 2 | 821 | 71 | ||
Set Membership: |
|