Search
Search

MYBPH| Gene Symbol: MYBPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268559.2 | XP_008266781.1 | 3 - 4 | 1107 | 83 | |
| XM_008268560.2 | XP_008266782.1 | 3 - 4 | 1109 | 83 | ||
Set Membership: |
|