Search
Search

ECHDC1| Gene Symbol: ECHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714807.3 | XP_002714853.1 | 4 - 5 | 511 | 88 | |
| XM_008263477.2 | XP_008261699.1 | 4 - 5 | 800 | 88 | ||
| XM_008263478.2 | XP_008261700.1 | 4 - 5 | 537 | 88 | ||
Set Membership: |
|