Search
Search

SMARCC2| Gene Symbol: SMARCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256642.1 | XP_008254864.1 | 21 - 22 | 2505 | 62 | |
| XM_017341933.1 | XP_017197422.1 | 20 - 21 | 2412 | 62 | ||
| XM_017341934.1 | XP_017197423.1 | 20 - 21 | 2409 | 62 | ||
| XM_017341935.1 | XP_017197424.1 | 21 - 22 | 2502 | 62 | ||
| XM_017341936.1 | XP_017197425.1 | 21 - 22 | 2505 | 62 | ||
| XM_017341937.1 | XP_017197426.1 | 20 - 21 | 2412 | 62 | ||
Set Membership: |
|