Search
Search

SEMA3B| Gene Symbol: SEMA3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713211.3 | XP_002713257.1 | 9 - 10 | 2143 | 52 | |
| XM_008260681.2 | XP_008258903.1 | 9 - 10 | 1189 | 52 | ||
| XM_008260682.2 | XP_008258904.1 | 9 - 10 | 1184 | 52 | ||
| XM_017343830.1 | XP_017199319.1 | 8 - 9 | 1262 | 52 | ||
| XM_017343831.1 | XP_017199320.1 | 10 - 11 | 1098 | 52 | ||
| XM_017343832.1 | XP_017199321.1 | 9 - 10 | 1095 | 52 | ||
| XM_017343833.1 | XP_017199322.1 | 4 - 5 | 466 | 52 | ||
Set Membership: |
|