Search
Search

PLEKHB1| Gene Symbol: PLEKHB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708706.3 | XP_002708752.1 | 5 - 6 | 666 | 67 | |
| XM_002708708.3 | XP_002708754.1 | 5 - 6 | 498 | 67 | ||
Set Membership: |
|