Search
Search
| Gene Symbol: VIT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008254542.2 | XP_008252764.1 | 15 - 16 | 2142 | 66 | |
| XM_008254543.2 | XP_008252765.1 | 15 - 16 | 2103 | 66 | ||
| XM_008254544.2 | XP_008252766.1 | 14 - 15 | 2063 | 66 | ||
| XM_008254546.2 | XP_008252768.1 | 14 - 15 | 2024 | 66 | ||
Set Membership: |
|