Search
Search

ZNF521| Gene Symbol: ZNF521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261154.2 | XP_008259376.1 | 6 - 7 | 3901 | 68 | |
| XM_017344089.1 | XP_017199578.1 | 5 - 6 | 3894 | 68 | ||
| XM_017344090.1 | XP_017199579.1 | 5 - 6 | 3883 | 68 | ||
Set Membership: |
|