Search
Search
| Gene Symbol: LPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266644.2 | XP_008264866.1 | 5 - 6 | 520 | 63 | |
| XM_017346981.1 | XP_017202470.1 | 5 - 6 | 519 | 63 | ||
| XM_017346982.1 | XP_017202471.1 | 4 - 5 | 487 | 63 | ||
| XM_017346983.1 | XP_017202472.1 | 4 - 5 | 500 | 63 | ||
| XM_017346984.1 | XP_017202473.1 | 4 - 5 | 382 | 63 | ||
Set Membership: |
|