Search
Search

RADIL| Gene Symbol: RADIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723067.3 | XP_002723113.3 | 3440 | 100 | ||
| XM_017339074.1 | XP_017194563.1 | 2576 | 100 | |||
| XM_017339075.1 | XP_017194564.1 | 2498 | 100 | |||
Set Membership: |
|