Search
Search

AASDH| Gene Symbol: AASDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717077.3 | XP_002717123.2 | 11 - 12 | 2355 | 91 | |
| XM_008267824.2 | XP_008266046.1 | 11 - 12 | 2355 | 91 | ||
| XM_008267825.2 | XP_008266047.1 | 11 - 12 | 2239 | 91 | ||
| XM_008267827.2 | XP_008266049.1 | 11 - 12 | 2182 | 91 | ||
| XM_008267829.2 | XP_008266051.1 | 10 - 11 | 2051 | 91 | ||
| XM_008267832.2 | XP_008266054.1 | 9 - 10 | 1469 | 91 | ||
| XM_017347539.1 | XP_017203028.1 | 11 - 12 | 2416 | 91 | ||
Set Membership: |
|