Search
Search

PHOSPHO1| Gene Symbol: PHOSPHO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719305.3 | XP_002719351.1 | 2 - 3 | 398 | 97 | |
| XM_008271327.2 | XP_008269549.1 | 2 - 3 | 400 | 97 | ||
| XM_008271330.2 | XP_008269552.1 | 2 - 3 | 395 | 97 | ||
Set Membership: |
|