Search
Search

WNK1| Gene Symbol: WNK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712781.3 | XP_002712827.1 | 21 - 22 | 6492 | 58 | |
| XM_008259830.2 | XP_008258052.1 | 21 - 22 | 6493 | 58 | ||
| XM_008259831.2 | XP_008258053.1 | 21 - 22 | 6490 | 58 | ||
| XM_008259832.2 | XP_008258054.1 | 21 - 22 | 6493 | 58 | ||
| XM_008259833.2 | XP_008258055.1 | 20 - 21 | 6409 | 58 | ||
| XM_008259834.2 | XP_008258056.1 | 21 - 22 | 6493 | 58 | ||
| XM_008259835.2 | XP_008258057.1 | 21 - 22 | 6489 | 58 | ||
| XM_008259837.2 | XP_008258059.1 | 20 - 21 | 6034 | 58 | ||
| XM_008259840.2 | XP_008258062.1 | 20 - 21 | 6031 | 58 | ||
| XM_008259841.2 | XP_008258063.1 | 20 - 21 | 6033 | 58 | ||
| XM_008259842.2 | XP_008258064.1 | 20 - 21 | 6030 | 58 | ||
| XM_008259843.2 | XP_008258065.1 | 19 - 20 | 5752 | 58 | ||
| XM_008259844.2 | XP_008258066.1 | 19 - 20 | 5751 | 58 | ||
| XM_008259845.2 | XP_008258067.1 | 19 - 20 | 5748 | 58 | ||
| XM_008259846.2 | XP_008258068.1 | 18 - 19 | 4525 | 58 | ||
| XM_008259847.2 | XP_008258069.1 | 18 - 19 | 4525 | 58 | ||
| XM_008259848.2 | XP_008258070.1 | 21 - 22 | 7275 | 58 | ||
| XM_017343442.1 | XP_017198931.1 | 20 - 21 | 6211 | 58 | ||
| XM_017343443.1 | XP_017198932.1 | 21 - 22 | 6494 | 58 | ||
Set Membership: |
|