Search
Search

LOC100355380| Gene Symbol: LOC100355380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714347.3 | XP_002714393.2 | 7 | 1185 | 62 | |
| XM_008262671.2 | XP_008260893.1 | 8 | 1215 | 62 | ||
Set Membership: |
|