Search
Search

SUPT7L| Gene Symbol: SUPT7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709948.3 | XP_002709994.1 | 2 - 3 | 348 | 63 | |
| XM_008254586.2 | XP_008252808.1 | 2 - 3 | 367 | 63 | ||
Set Membership: |
|