Search
Search

NOL11| Gene Symbol: NOL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271729.2 | XP_008269951.1 | 10 - 11 | 1191 | 71 | |
| XM_008271730.2 | XP_008269952.1 | 10 - 11 | 1188 | 71 | ||
Set Membership: |
|