Search
Search

SLC6A9| Gene Symbol: SLC6A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715153.3 | XP_002715199.1 | 6 - 7 | 974 | 56 | |
| XM_017346366.1 | XP_017201855.1 | 7 - 8 | 965 | 56 | ||
| XM_017346367.1 | XP_017201856.1 | 9 - 10 | 1263 | 56 | ||
| XM_017346368.1 | XP_017201857.1 | 5 - 6 | 758 | 56 | ||
| XM_017346369.1 | XP_017201858.1 | 4 - 5 | 624 | 56 | ||
Set Membership: |
|