Search
Search

YEATS2| Gene Symbol: YEATS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716492.3 | XP_002716538.2 | 20 - 21 | 3180 | 59 | |
| XM_008266625.2 | XP_008264847.1 | 19 - 20 | 2942 | 59 | ||
| XM_017346973.1 | XP_017202462.1 | 20 - 21 | 2989 | 59 | ||
Set Membership: |
|