Search
Search

RAB23| Gene Symbol: RAB23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714520.3 | XP_002714566.1 | 2 - 3 | 510 | 122 | |
| XM_017345219.1 | XP_017200708.1 | 2 - 3 | 515 | 122 | ||
| XM_017345220.1 | XP_017200709.1 | 2 - 3 | 513 | 122 | ||
Set Membership: |
|