Search
Search

C14H21orf58| Gene Symbol: C14H21orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267244.2 | XP_008265466.1 | 7 - 8 | 873 | 94 | |
| XM_017347234.1 | XP_017202723.1 | 7 - 8 | 1176 | 94 | ||
Set Membership: |
|