Search
Search

LRCH3| Gene Symbol: LRCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266742.2 | XP_008264964.1 | 18 - 19 | 2073 | 65 | |
| XM_008266743.2 | XP_008264965.1 | 18 - 19 | 2070 | 65 | ||
| XM_008266745.2 | XP_008264967.1 | 17 - 18 | 2019 | 65 | ||
| XM_008266746.2 | XP_008264968.1 | 18 - 19 | 2072 | 65 | ||
| XM_008266747.2 | XP_008264969.1 | 18 - 19 | 1989 | 65 | ||
| XM_008266748.2 | XP_008264970.1 | 18 - 19 | 1986 | 65 | ||
| XM_008266749.2 | XP_008264971.1 | 17 - 18 | 1965 | 65 | ||
| XM_008266750.2 | XP_008264972.1 | 18 - 19 | 1989 | 65 | ||
| XM_008266751.2 | XP_008264973.1 | 16 - 17 | 1911 | 65 | ||
| XM_008266752.2 | XP_008264974.1 | 17 - 18 | 1965 | 65 | ||
| XM_017347035.1 | XP_017202524.1 | 16 - 17 | 1965 | 65 | ||
| XM_017347036.1 | XP_017202525.1 | 19 - 20 | 2163 | 65 | ||
| XM_017347037.1 | XP_017202526.1 | 19 - 20 | 2163 | 65 | ||
Set Membership: |
|