Search
Search

METTL15| Gene Symbol: METTL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269179.2 | XP_008267401.1 | 3 - 4 | 614 | 63 | |
| XM_008269197.2 | XP_008267419.1 | 4 - 5 | 720 | 63 | ||
| XM_017345657.1 | XP_017201146.1 | 3 - 4 | 614 | 63 | ||
| XM_017345662.1 | XP_017201151.1 | 3 - 4 | 614 | 63 | ||
Set Membership: |
|