Search
Search

CDC25A| Gene Symbol: CDC25A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260577.2 | XP_008258799.1 | 3 - 4 | 693 | 69 | |
| XM_017343797.1 | XP_017199286.1 | 3 - 4 | 693 | 69 | ||
Set Membership: |
|