Search
Search

IL6ST| Gene Symbol: IL6ST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262226.2 | XP_008260448.1 | 12 - 13 | 1830 | 77 | |
| XM_017344725.1 | XP_017200214.1 | 11 - 12 | 1744 | 77 | ||
| XM_017344726.1 | XP_017200215.1 | 11 - 12 | 1589 | 77 | ||
Set Membership: |
|