Search
Search

IL6ST| Gene Symbol: IL6ST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262226.2 | XP_008260448.1 | 13 - 14 | 1970 | 107 | |
| XM_017344725.1 | XP_017200214.1 | 12 - 13 | 1884 | 107 | ||
| XM_017344726.1 | XP_017200215.1 | 12 - 13 | 1729 | 107 | ||
Set Membership: |
|