Search
Search

DTWD1| Gene Symbol: DTWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717731.3 | XP_002717777.1 | 4 - 5 | 705 | 134 | |
| XM_008269038.2 | XP_008267260.1 | 3 - 4 | 534 | 134 | ||
| XM_017348035.1 | XP_017203524.1 | 4 - 5 | 678 | 134 | ||
Set Membership: |
|