Search
Search

MCUB| Gene Symbol: MCUB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267492.2 | XP_008265714.1 | 3 - 4 | 645 | 58 | |
| XM_017347350.1 | XP_017202839.1 | 4 - 5 | 702 | 58 | ||
| XM_017347351.1 | XP_017202840.1 | 4 - 5 | 699 | 58 | ||
Set Membership: |
|