Search
Search

RUSC2| Gene Symbol: RUSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002707928.3 | XP_002707974.2 | 4 - 5 | 3337 | 62 | |
| XM_008269824.2 | XP_008268046.1 | 4 - 5 | 3341 | 62 | ||
| XM_008269874.2 | XP_008268096.1 | 4 - 5 | 2936 | 62 | ||
Set Membership: |
|