Search
Search

KIAA0408| Gene Symbol: KIAA0408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714808.3 | XP_002714854.1 | 3 - 4 | 593 | 76 | |
| XM_017345359.1 | XP_017200848.1 | 3 - 4 | 1291 | 76 | ||
Set Membership: |
|