Search
Search

NOD2| Gene Symbol: NOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275042.2 | XP_008273264.1 | 2573 | 59 | ||
| XM_008275044.2 | XP_008273266.1 | 2573 | 59 | |||
| XM_017350884.1 | XP_017206373.1 | 2573 | 59 | |||
| XM_017350886.1 | XP_017206375.1 | 2078 | 59 | |||
Set Membership: |
|