Search
Search

KLC1| Gene Symbol: KLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248627.2 | XP_008246849.1 | 1944 | 72 | ||
| XM_008248631.2 | XP_008246853.1 | 1877 | 72 | |||
| XM_008248632.2 | XP_008246854.1 | 1850 | 72 | |||
Set Membership: |
|