Search
Search

VIPAS39| Gene Symbol: VIPAS39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719626.3 | XP_002719672.1 | 7 - 8 | 668 | 60 | |
| XM_008271985.2 | XP_008270207.1 | 6 - 7 | 522 | 60 | ||
Set Membership: |
|