Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Oc06798632_s1
          See other CELF4 GE Assays ›
          Gene Symbol:
          CELF4
          Gene Name:
          CUGBP Elav-like family member 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 78433543 - 78702239 on Build OryCun2.0
          Species:
          Rabbit
          Species Specific ID (Flybase ID):
          -
          Assay ID Oc06798632_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CELF4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008261235.2 XP_008259457.1 13 3548 96
          XM_008261236.2 XP_008259458.1 13 3548 96
          XM_008261237.2 XP_008259459.1 13 3521 96
          XM_008261238.2 XP_008259460.1 13 3518 96
          XM_008261239.2 XP_008259461.1 13 3494 96
          XM_008261240.2 XP_008259462.1 13 3491 96
          XM_008261241.2 XP_008259463.1 13 3488 96
          XM_008261242.2 XP_008259464.1 13 3488 96
          XM_008261243.2 XP_008259465.1 12 3407 96
          XM_008261244.2 XP_008259466.1 12 3407 96
          XM_008261246.2 XP_008259468.1 12 3380 96
          XM_008261247.2 XP_008259469.1 13 3491 96
          XM_008261248.2 XP_008259470.1 13 3515 96
          XM_017344123.1 XP_017199612.1 15 3652 96
          XM_017344124.1 XP_017199613.1 15 3652 96
          XM_017344125.1 XP_017199614.1 15 3652 96
          XM_017344126.1 XP_017199615.1 15 3652 96
          XM_017344127.1 XP_017199616.1 15 3649 96
          XM_017344128.1 XP_017199617.1 14 3570 96
          XM_017344129.1 XP_017199618.1 15 3625 96
          XM_017344130.1 XP_017199619.1 15 3622 96
          XM_017344131.1 XP_017199620.1 15 3619 96
          XM_017344132.1 XP_017199621.1 15 3616 96
          XM_017344133.1 XP_017199622.1 13 3554 96
          XM_017344135.1 XP_017199624.1 13 3488 96
          XM_017344136.1 XP_017199625.1 12 3410 96
          XM_017344137.1 XP_017199626.1 15 3743 96
          XM_017344138.1 XP_017199627.1 13 3458 96

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline