Search
Search

CEP83| Gene Symbol: CEP83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256887.2 | XP_008255109.1 | 14 - 15 | 2353 | 99 | |
| XM_008256888.2 | XP_008255110.1 | 15 - 16 | 2258 | 99 | ||
| XM_008256890.2 | XP_008255112.1 | 12 - 13 | 1594 | 99 | ||
| XM_017342022.1 | XP_017197511.1 | 15 - 16 | 2619 | 99 | ||
Set Membership: |
|