Search
Search

FHL1| Gene Symbol: FHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720323.3 | XP_002720369.1 | 2 - 3 | 343 | 60 | |
| XM_008273250.2 | XP_008271472.2 | 3 - 4 | 273 | 60 | ||
| XM_008273251.2 | XP_008271473.2 | 2 - 3 | 398 | 60 | ||
| XM_008273253.2 | XP_008271475.1 | 2 - 3 | 368 | 60 | ||
| XM_017349780.1 | XP_017205269.1 | 3 - 4 | 344 | 60 | ||
| XM_017349781.1 | XP_017205270.1 | 2 - 3 | 350 | 60 | ||
Set Membership: |
|