Search
Search

LOC4325473| Gene Symbol: LOC4325473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760918.2 | XP_015616404.1 | 1 - 2 | 1297 | 70 | |
| GenBank mRNA | AK065080.1 | - | 1 - 2 | 1280 | 70 | |
| AK070687.1 | - | 1 - 2 | 224 | 70 | ||
| AK098852.1 | - | 1 - 2 | 1249 | 70 | ||
| AK098894.1 | - | 1 - 2 | 1247 | 70 | ||
| AK099060.1 | - | 1 - 2 | 1248 | 70 | ||
Set Membership: |
|