Search
Search

LOC4325038| Gene Symbol: LOC4325038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758367.1 | XP_015613853.1 | 4 - 5 | 1147 | 98 | |
| GenBank mRNA | AK068254.1 | - | 4 - 5 | 1137 | 98 | |
| AK105713.1 | - | 4 - 5 | 1153 | 98 | ||
| HQ858859.1 | - | 4 - 5 | 1038 | 98 | ||
| HQ858868.1 | - | 4 - 5 | 1038 | 98 | ||
Set Membership: |
|