Search
Search

LOC4333494| Gene Symbol: LOC4333494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773307.2 | XP_015628793.1 | 4 - 5 | 1173 | 94 | |
| GenBank mRNA | AK066101.1 | - | 4 - 5 | 1001 | 94 | |
| AK099619.1 | - | 4 - 5 | 1001 | 94 | ||
Set Membership: |
|