Search
Search

LOC4337605| Gene Symbol: LOC4337605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784097.2 | XP_015639583.1 | 2 - 3 | 437 | 81 | |
| GenBank mRNA | AK102177.1 | - | 1 - 2 | 2068 | 81 | |
Set Membership: |
|