Search
Search

LOC4342588| Gene Symbol: LOC4342588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789236.2 | XP_015644722.1 | 1 - 2 | 651 | 79 | |
| GenBank mRNA | AK071133.1 | - | 1 - 2 | 651 | 79 | |
Set Membership: |
|