Search
Search

LOC4346315| Gene Symbol: LOC4346315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794260.1 | XP_015649746.1 | 6 - 7 | 1235 | 68 | |
| GenBank mRNA | AK065854.1 | - | 6 - 7 | 1238 | 68 | |
Set Membership: |
|